Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-APC Clone: CC-1,

Catalog No. OP80100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
OP80100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OP80100UG Supplier MilliporeSigma™ Supplier No. OP80100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects APC Clone: CC-1 in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Immunofluorescence

Germ line mutations in the APC gene are responsible for familial adenomatous polyposis and somatic mutations contribute to sporadic colorectal cancer. The 312kDa APC protein is expressed in all tissues and localized to the cytoskeletal architecture in an insoluble complex with β-catenin. The APC (Ab-7) mouse monoclonal antibody clone CC-1 decorates oligodendrocyte cell bodies, purkinje cells, astrocytes, and optic nerve cell bodies of formalin-fixed, paraffin-embedded, or fresh frozen sections from human, mouse, and rat brain. Dectection of APC (Ab-7) can be perfomed using immunofluorescence or immunohistochemistry techniques.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC
Applications Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone CC-1
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen a recombinant protein consisting of amino acids 1-226 of APC
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes APC in oligodendrocytes and astrocytes. Antibody Target Gene Symbol: APC Target Synonym: AI047805, Apc7, AU020952, AW124434, BTPS2, DP2, DP2.5, DP3, Familial adenomatous polyposis, FAP, GS, Min, RATAPC Entrez Gene Name: adenomatous polyposis coli
Target Species Human, Mouse, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG2b
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.