Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc11 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP190139
Description
Apc11 Polyclonal specifically detects Apc11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Apc11 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| anaphase promoting complex subunit 11, anaphase promoting complex subunit 11 (yeast APC11 homolog), anaphase-promoting complex subunit 11, APC11 anaphase promoting complex subunit 11 homolog, APC11Hepatocellular carcinoma-associated RING finger protein, Apc11p, Cyclosome subunit 11, HSPC214, MGC882 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ANAPC11 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ANDENCGICRMAFNGCCPDCPLHGESISRCLGWCPQPVPVLGGRAHPQVPINTASPTPGQHTGSLMSREESSRSPDPTPPAL | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 51529 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction