Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC2 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA130266
Description
PA1-30272 detects PRMT2 from human samples. PA1-30272 has been successfully used in Chimmunoprecipitation, ELISA, Western blot applications. The PA1-30272 immunogen is: Synthetic peptide: AIADYAATDETQLSFLR, corresponding to amino acids 37-53 of Human PR
A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the
Specifications
APC2 | |
Polyclonal | |
APC2 | |
APCL | |
Rabbit | |
Protein A | |
RUO | |
10297, 23805 | |
4° C, do not freeze | |
Liquid |
Western Blot | |
Unconjugated | |
O95996, Q9Z1K7 | |
APC2 | |
Synthetic peptide, corresponding to amino acids 810-822 of Human Apc2. | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction