Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apelin Receptor, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA521285
Description
PA5-21285 targets Apelin receptor in IHC (P), ICC, IF and WB applications and shows reactivity with Human samples.The PA5-21285 immunogen is a synthetic peptide corresponding to a region within amino acids 2 and 96 of Apelin Receptor (Uniprot ID#P35414).
Specifications
Apelin receptor | |
Polyclonal | |
Unconjugated | |
P35414 | |
APLNR | |
Synthetic peptide corresponding to a region within amino acids 2 and 96 of Apelin Receptor (Uniprot ID#P35414) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1mg/mL | |
APLNR | |
AGTRL1, APJ, APJR, HG11, APJ (apelin) receptor, APJ receptor, G protein-coupled receptor APJ, G-protein coupled receptor APJ, G-protein coupled receptor HG11, HG11 orphan receptor, angiotensin II receptor-like 1, angiotensin receptor-like 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
187 | |
Store at -20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction