Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

apelin, Mouse, Clone: 1A8-2A5, Abnova™

Catalog No. 89001107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-107 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-107 Supplier Abnova Corporation Supplier No. H00008862M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant APLN.

Apelin is a neuropeptide expressed in the supraoptic and paraventricular nuclei that acts on specific receptors located on vasopressinergic neurons (De Mota et al., 2004) [PubMed 15231996].[supplied by OMIM

Sequence: MSATWCSPEGQGMGQGPGREVGGNSAASGPASPIRDPCLSEAGLKGPPSAHPRRLCLLHRLVCFSGGLTSIQLSPRTCCSHQWAQLFSPACFPQWRAPGCSLDDSRSLTRIRPVHLPGPSLD

Specifications

Antigen apelin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A8-2A5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant APLN.
Formulation PBS with no preservative; pH 7.4
Gene APLN
Gene Accession No. BC021104.1
Gene Alias XNPEP2
Gene Symbols APLN
Host Species Mouse
Immunogen APLN (AAH21104.1, 1 a.a. ∼ 122 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 8862
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.