Learn More
apelin, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human APLN protein.
Supplier: Abnova Corporation H00008862B01
Description
Apelin is a neuropeptide expressed in the supraoptic and paraventricular nuclei that acts on specific receptors located on vasopressinergic neurons (De Mota et al., 2004) [PubMed 15231996].[supplied by OMIM
Sequence: MSATWCSPEGQGMGQGPGREVGGNSAASGPASPIRDPCLSEAGLKGPPSAHPRRLCLLHRLVCFSGGLTSIQLSPRTCCSHQWAQLFSPACFPQWRAPGCSLDDSRSLTRIRPVHLPGPSLDSpecifications
apelin | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length human APLN protein. | |
APLN | |
XNPEP2 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
Western Blot | |
Unconjugated | |
No additive | |
BC021104 | |
APLN | |
APLN (AAH21104.1, 1 a.a. ∼ 122 a.a) full-length human protein. | |
RUO | |
Yes | |
8862 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.