Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Apobec3 Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. 07723MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-723-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-723-MI Supplier MilliporeSigma Supplier No. 07723

Rabbit Polyclonal Antibody

Anti-Apobec3 Antibody, NT is a high quality Rabbit Polyclonal Antibody for the detection of Apobec3 and has been validated in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apobec3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified antibody in 0.1M Tris-Glycine, 0.15M NaCl, 0.05% Sodium Azide, pH 7.4.
Gene Symbols APOBEC3F; MGC74891; KA6; BK150C2.4.MRNA; ARP8
Host Species Rabbit
Immunogen KLH-conjugated, synthetic peptide corresponding to amino acids 82-95 (C-DKVLKVLSPREEFK) of mouse apolipoprotein B editing complex 3 (Apobec3), with an N-terminal cysteine added for conjugation purposes.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001006666.1; NM_145298.5
Target Species Mouse
Content And Storage 2-8°C for 12 months
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.