Learn More
APOBEC3A, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human APOBEC3A peptide using ARM Technology.
Supplier: Abnova Corporation H00200315K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. This gene encodes a protein that lacks the zinc binding activity and may be an expressed pseudogene. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
APOBEC3A | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
APOBEC3A | |
APOBEC3A | |
A synthetic peptide of human APOBEC3A | |
RUO | |
200315 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
apolipoprotein B mRNA editing enzyme, catalytic polypeptide-like 3A | |
APOBEC3A | |
ARP3, PHRBN, bK150C2.1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.