Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-ApoE4 Fragment nApoECF, Polyclonal

Catalog No. ABN1350MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1350MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1350MI Supplier MilliporeSigma™ Supplier No. ABN1350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects ApoE4 Fragment nApoECF Clone: in Human samples, and it is validated for Immunofluorescence, Immunohistochemistry (Paraffin), Western Blotting

Apolipoprotein E (UniProt P02649; also known as Apo-E) is encoded by the APOE (also known as AD2, LDLCQ5, LPG) gene (Gene ID 348) in human. Apolipoprotein E (ApoE) is found in the chylomicron and Intermediate-density lipoprotein (IDLs) and is essential for the normal catabolism of triglyceride-rich lipoprotein constituents. ApoE is initially produced with a signal peptide sequence (a.a. 1-18), the removal of which yields the 299 a.a. mature protein. Three additional natural variants (E2, E3, and E4) exist due to polymorphisms. E4 has an allele frequency of approximately 14 percent and has been implicated in atherosclerosis and various CNS disorders, including Alzheimer's disease (AD). ApoE4 is highly susceptible to proteolysis, which impairs its role in cholesterol transport and beta-amyloid removal in the CNS. Various ApoE4 fragments (14–20kDa) have been identified in the AD brain. The C-terminal portion of apoE has been implicated in binding to beta-amyloid and is localized to plaques, while the N-terminal domain preferentially localizes within neurofibrillary tangles (NFTs) in the AD brain.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ApoE4 Fragment nApoECF
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P02649
Gene Symbols APOE
Host Species Rabbit
Immunogen Linear peptide corresponding to the C-terminal sequence of human ApoE4 fragment nApoECF (Asp172).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000032
Test Specificity Specifically recognizes the ApoE4 N-terminal fragment(s) generated by a proteolytic cleavage between Asp172 and Leu173 of ApoE4. Does not recognize full-length ApoE3 and ApoE4, nor ApoE4 fragments with C-termi other than D172.
Target Species Human
Content And Storage 2°C to 8°C for one year
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.