Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOF, Mouse, Clone: 1D5, Abnova™

Catalog No. 89016161 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-161 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-161 Supplier Abnova Corporation Supplier No. H00000319M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant APOF.

The product of this gene is one of the minor apolipoproteins found in plasma. This protein forms complexes with lipoproteins and may be involved in transport and/or esterification of cholesterol. [provided by RefSeq

Sequence: TSYGKQTNVLMHFPLSLESQTPSSDPLSCQFLHPKSLPGFSHMAPLPKFLVSLALRNDALEEAGCQADVWALQLQLYRQGGVNATQVLIQHLRGLQKGRSTERNVSVEALASALQLLAREQQSTGRVGRSLPTEDCENEKEQAVHNVVQLLPGVGTFYNLGTALYYATQNCLGKARERGRDGAIDLGYDLLMTMAGMSGGPMGLAISAALKPALRSGVQQLIQYYQDQKDANISQPETTKEGLRAISDVSDLEETTTLASFISEVVSSAPYWGWAIIKSYDLDPGAGSLEI

Specifications

Antigen APOF
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant APOF.
Formulation PBS with no preservative; pH 7.4
Gene APOF
Gene Accession No. BC026257
Gene Alias Apo-F/DKFZp781G18150/LTIP/MGC22520
Gene Symbols APOF
Host Species Mouse
Immunogen APOF (AAH26257, 37 a.a. ∼ 327 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.