Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

apolipoprotein H (beta-2-glycoprotein I), Mouse, Clone: 3F10, Abnova™

Catalog No. 89001425 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-425 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-425 Supplier Abnova Corporation Supplier No. H00000350M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant APOH.

Apolipoprotein H has been implicated in a variety of physiologic pathways including lipoprotein metabolism, coagulation, and the production of antiphospholipid autoantibodies. APOH may be a required cofactor for anionic phospholipid binding by the antiphospholipid autoantibodies found in sera of many patients with lupus and primary antiphospholipid syndrome, but it does not seem to be required for the reactivity of antiphospholipid autoantibodies associated with infections. [provided by RefSeq

Sequence: DGYSLDGPEEIECTKLGNWSAMPSCKASCKVPVKKATVVYQGERVKIQEKFKNGMLHGDKVSFFCKNKEKKCSYTEDAQCIDGTIEVPKCFKEHSSLAFWKTDASDVKPC

Specifications

Antigen apolipoprotein H (beta-2-glycoprotein I)
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3F10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant APOH.
Formulation ascites with no preservative
Gene APOH
Gene Accession No. NM_000042
Gene Alias B2G1/BG
Gene Symbols APOH
Host Species Mouse
Immunogen APOH (NP_000033, 236 a.a. ∼ 345 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 350
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.