Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOL4, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017173 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-173 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-173 Supplier Abnova Corporation Supplier No. H00080832B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human APOL4 protein.

The protein encoded by this gene is a member of the apolipoprotein L family and may play a role in lipid exchange and transport throughout the body, as well as in reverse cholesterol transport from peripheral cells to the liver. Two transcript variants encoding two different isoforms have been found for this gene. Only one of the isoforms appears to be a secreted protein. [provided by RefSeq

Sequence: MGSWVQLITSVGTSGLFLGVRVREEGAGMRCSKTIQAGQWLDSSKGPLGPSPPPVPTAGYSSSFCVHYVNLLPGVLVLSVTSQYPHLSMALCQLAAHDWPRLSCVCV

Specifications

Antigen APOL4
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human APOL4 protein.
Formulation PBS with no preservative; pH 7.4
Gene APOL4
Gene Accession No. ENST00000328429
Gene Alias APOL-IV/APOLIV
Gene Symbols APOL4
Host Species Mouse
Immunogen APOL4 (ENSP00000331089, 1 a.a. ∼ 107 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80832
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.