Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Apolipoprotein A-I, Mouse, Unlabeled, Clone: A/13,

Catalog No. MAB011A13 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
250 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB011A13 250 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB011A13 Supplier MilliporeSigma™ Supplier No. MAB011A/13
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein A-I
Applications Radioimmune Assays (RIA)
Classification Monoclonal
Clone A/13
Concentration Please refer to the Certificate of Analysis for the lot-specific concentration.
Conjugate Unconjugated
Formulation IgG fraction purified from mouse ascites by DEAE column chromatography. In 0.015M potassium phosphate buffer with 0.15M NaCl, 0.1% Sodium Azide, pH 7.2.
Gene APOA1, ApoA-I, preproapolipoprotein, amyloidosis, MGC117399, Apo-AI
Gene Accession No. P02647
Gene Symbols APOA1, ApoA-I, preproapolipoprotein, amyloidosis, MGC117399, Apo-AI
Host Species Mouse
Immunogen Purified human plasma Apo A-I.
Quantity 250 μg
Regulatory Status RUO
Research Discipline Lipid Metabolism and Weight Regulation
Primary or Secondary Primary
Gene ID (Entrez) NM_000039.1
Test Specificity Apolipoprotein A-I. No cross reactivity to Apo A-II or Apo B. Affinity Constant: 8 x 109L/mol
Target Species Human
Content And Storage Maintain at -20°C in undiluted aliquots for up to 12 months. Do not store below -20°C or in dry ice as precipitation may occur. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.