Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Apolipoprotein C-III, Mouse, Unlabeled, Clone: 68/7,

Catalog No. MAB002687 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB002687 500 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB002687 Supplier MilliporeSigma™ Supplier No. MAB00268/7

Mouse Monoclonal Antibody

Apolipoprotein C-III is a very low density lipoprotein (VLDL) protein. APOC3 inhibits lipoprotein lipase and hepatic lipase; it is thought to delay catabolism of triglyceride-rich particles. The APOA1, APOC3 and APOA4 genes are closely linked in both rat and human genomes. The A-I and A-IV genes are transcribed from the same strand, while the A-1 and C-III genes are convergently transcribed. An increase in apoC-III levels induces the development of hypertriglyceridemia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein C-III
Applications Radioimmune Assays (RIA)
Classification Monoclonal
Clone 68/7
Concentration Please refer to the Certificate of Analysis for the lot-specific concentration.
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in 0.02M Phosphate buffer,PH 7.6 0.25M NaCl with 0.1% Sodium Azide.
Gene APOC3, ApoC-III, Apo-CIII, APOCIII, MGC150353
Gene Accession No. P02656
Gene Symbols APOC3, ApoC-III, Apo-CIII, APOCIII, MGC150353
Host Species Mouse
Quantity 500 μg
Regulatory Status RUO
Research Discipline Lipid Metabolism and Weight Regulation
Primary or Secondary Primary
Gene ID (Entrez) NM_000040.1
Test Specificity Apolipoprotein C-III. No reactivity to HDL, A2, VLDL, C2, LDL or AI.
Target Species Human
Content And Storage Maintain at 2°-8°C in undiluted aliquots for up to six months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.