Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Apolipoprotein E, Polyclonal

Catalog No. 178479500UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-847-9500UL 500 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-847-9500UL Supplier MilliporeSigma™ Supplier No. 178479500UL
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Specifically detects Apolipoprotein E Clone: in Human, Mouse, Rat samples, and it is validated for ELISA, Immunoprecipitation

TRUSTED_SUSTAINABILITY

Specifications

Antigen Apolipoprotein E
Applications ELISA, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation In 500mM NaCl, 50mM Tris-HCl, pH 7.5.
Host Species Goat
Immunogen purified recombinant human apolipoprotein E
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes human apolipoprotein E. Does not cross-react with other apolipoproteins. Antibody Target Gene Symbol: APOE Target Synonym: AD2, AI255918, APOE2, APOE4, APOEA, APOLIPOPROTEIN E, LDLCQ5, LPG, MGC1571 Entrez Gene Name: apolipoprotein E
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Diluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.