Learn More
Description
Specifications
Specifications
| Antigen | Apolipoprotein J |
| Applications | ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.125M Sodium Borate buffer, 0.075M Sodium Chloride, 0.005M EDTA, pH 8.0, with 0.01% sodium azide. |
| Gene Symbols | CLU; SP-40; MGC24903; NA1/NA2; APOJ; CLI; SGP-2; TRPM-2; AAG4; TRPM2; KUB1; Apo-J; SGP2 |
| Host Species | Goat |
| Immunogen | Apolipoprotein J, purified from human plasma. |
| Purification Method | Affinity Purified |
| Quantity | 500 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.