Learn More
Description
The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized. [provided by RefSeq
Specifications
Specifications
| Antigen | apolipoprotein M |
| Applications | ELISA, KnockDown, Western Blot |
| Classification | Monoclonal |
| Clone | 1F10 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant APOM. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | APOM |
| Gene Accession No. | BC020683 |
| Gene Alias | G3a/HSPC336/MGC22400/NG20 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.