Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

apolipoprotein M, Mouse, Clone: 2E5, Abnova™

Catalog No. 89001318 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-318 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-318 Supplier Abnova Corporation Supplier No. H00055937M02
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant APOM.

The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized. [provided by RefSeq

Sequence: CPEHSQLTTLGVDGKEFPEVHLGQWYFIAGAAPTKEELATFDPVDNIVFNMAAGSAPMQLHLRATIRMKDGLCVPRKWIYHLTEGSTDLRTEGRPDMKTELFSSSCPGGIMLNETGQGYQRFLLYNRSPHPPEKCVEEFKSLTSCLDSKAFLLTPRNQEACELSNN

Specifications

Antigen apolipoprotein M
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant APOM.
Formulation PBS with no preservative; pH 7.4
Gene APOM
Gene Accession No. BC020683
Gene Alias G3a/HSPC336/MGC22400/NG20
Gene Symbols APOM
Host Species Mouse
Immunogen APOM (AAH20683, 23 a.a. ∼ 188 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55937
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.