Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ anti-APP A4, a.a. 66-81 of APP {NT} Alexa Fluor 647, Clone: 22C11,
Mouse Monoclonal Antibody
Supplier: MilliporeSigma™ MAB348AF647
Description
Specifically detects APP A4, a.a. 66-81 of APP {NT} Clone: 22C11 in Canine, Fish, Human, Monkey, Mouse, Porcine, Rat samples, and it is validated for Immunocytochemistry
Amyloid beta A4 protein (UniProt P05067; also known as ABPP, Alzheimer disease amyloid protein, Amyloid precursor protein, APP, APPI, Cerebral vascular amyloid peptide, CVAP, PN-II, PreA4, Protease nexin-II) is encoded by the APP (also known as A4, AD1) gene (Gene ID 351) in human. Amyloid precursor protein (APP) is initially produced with a signal peptide sequence (a.a. 1-17), the removal of which yields the mature protein with a large extracellular portion (a.a. 18-699), followed by a transmembrane segment (a.a. 700-723) and a cytoplasmic (a.a. 724-770) tail. APP can be further processed by the α-, β-, and γ-secretases in two alternative processing pathways. In the non-amyloidogenic pathway, APP is first cleaved by the plasma membrane-localized α-secretase to generate an N-terminal extracellular sAPPα fragment (a.a. 18-687) and a membrane-bound C-terminal fragment C83 (CTFα), which can be further cleaved by γ-secretase to produce a non-toxic small peptide p3 and a cytoplasmic APP intracellular domain (AICD). In the amyloidogenic pathway, APP undergoes β-cleavage in BACE-1 (β-site APP-cleaving enzyme)-enriched endosomes to generate an N-terminal extracellular sAPPβ fragment (a.a. 18-671) and a membrane-bound C-terminal fragment C99 (CTFβ). Subsequent cleavage of C99 by γ-secretase releases the amyloid β peptide (Aβ) and AICD. Aβ accumulation in the cortical and hippocampal regions of the brain is a major pathological feature of Alzheimer's disease (AD).
Specifications
APP A4, a.a. 66-81 of APP {NT} | |
Monoclonal | |
Alexa Fluor 647 | |
P05067 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Clone 22C11 reacts with pre-A4. The antibody recognizes amino acids 66-81 of the N-terminus on the pre-A4 molecule (Hilbich, C., et al. (1993). J. Biol. Chem. 268(35):26571-26577). Clone 22C11 recognizes all three isoforms of APP, immature ∽110kDa, sAPP ∽120kDa, and mature ∽130kDa (Hoffmann, J., et al. (2000). Eur. J. Cell. Biol.79(12):905-914.). The antibody is known to cross react with APLP2 (Slunt, H.H., et al. (1994). J. Biol. Chem. 269(4):2637-2644). | |
2°C to 8°C for one year from date of shipment | |
IgG1 |
Immunocytochemistry | |
22C11 | |
Purified mouse monoclonal IgG1 antibody conjugate in PBS with 15mg/mL BSA and 0.1 % sodium azide. | |
APP, A4, AD1 | |
Purified recombinant Alzheimer precursor A4 (pre-A4695) fusion protein. | |
100 μL | |
Neuroscience | |
NM_201414, NM_000484.2, NM_201413.1 | |
Canine, Fish, Human, Monkey, Mouse, Pig, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction