Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP A4 Mouse anti-Canine, Human, Mouse, Monkey, Pig, Rat, Fish, Clone: 22C11, MilliporeSigma™

Catalog No. IHCR10026MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
6 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IHCR10026MI 6 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IHCR10026MI Supplier MilliporeSigma Supplier No. IHCR10026
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Alzheimer Precursor Protein A4 using this Anti-APP A4 Antibody, a.a. 66-81 (N-terminus), Prediluted, clone 22C11 validated for use in immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen APP A4
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 22C11
Conjugate Unconjugated
Formulation Liquid diluted in PBS, pH 7.2 with stabilizers, 0.2% Tween 20, and 0.1% ProClin 300 as preservative.
Gene Accession No. NM_000484.2; NM_201413.1; NM_201414.1
Gene Symbols APP, ABPP, PN-II, AD1, A4, CVAP, PN2, ABETA, CTFgamma, APPI, PreA4, AAA
Host Species Mouse
Immunogen Purified recombinant Alzheimer precursor A4 (pre A4695) fusion protein.
Quantity 6 mL
Primary or Secondary Primary
Target Species Canine, Fish, Human, Monkey, Mouse, Pig, Rat
Content And Storage 2-8°C.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.