Learn More
Description
Amyloid precursor protein (APP) gene encodes multiple APPs ranging from 695 to 770 amino acids. These proteins are processed into β-Amyloid peptides (39-43 amino acids), which form abnormal plaques in the cerebral cortex and blood vessel walls during Alzheimer's disease. The transmembrane form of APP is a putative cell surface receptor that possesses neurite-promoting activity, co-localizes with the plaques found in Alzheimer's disease, and is involved in synaptic vesicle recycling. APP binding protein 1 (APP-BP1) interacts with the C-terminus of APP, and is a relative of the ubiquitin-activating enzymes (E1). The structure of APP-BP1 includes a human Uba3 binding site (UBS) at amino acids 443 to 479. This site may be important for APP-BP1 regulation of the cell cycle through interactions with ubiquitinylation-related pathways. Transfection of APP-BP1 in cells with a ts41 mutation suppresses the abnormal S-phases observed in these cells in a hUba3- and hUbc12-dependent manner. In addition, APP-BP1 has been implicated in ubiquitinylation-dependent apoptosis in neurons. Thus, APP-BP1 may be a multi-functional APP-binding protein that regulates cell cycle dynamics.
Host Species: Mouse
Clone: 20
Isotype: IgG2a
Species Reactivity [for Features Main]: Mouse
Immunogen: Human APP-BP1 aa. 348-464
Immunofluorescence, Western Blotting
Specifications
Specifications
| Antigen | APP-BP1 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 20 |
| Concentration | 250μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Human APP-BP1 aa. 348-464 |
| Purification Method | Affinity Purified |
| Quantity | 150 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.