Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APP Mouse anti-Human, Rat, Clone: 2.F2.19B4, MilliporeSigma™

Catalog No. MAB343MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB343MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB343MI Supplier MilliporeSigma Supplier No. MAB343
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-APP Antibody, APP 643-695 C-terminal fragment, clone 2.F2.19B4 is validated for use in immunohistochemistry (paraffin), western blotting for the detection of Alzheimer Precursor Protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 2.F2.19B4
Conjugate Unconjugated
Formulation Liquid.
Gene Accession No. NM_000484.2; NM_201413.1; NM_201414.1
Gene Symbols APP, PN-II, AD1, ABETA, CTFgamma, APPI, ABPP, A4, CVAP, PN2, PreA4, AAA
Host Species Mouse
Immunogen Carboxyl fragment of APP 643-695 / Jonas.
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.