Learn More
Description
Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. [provided by RefSeq
Specifications
Specifications
| Antigen | AQP8 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 1A1 |
| Conjugate | Unconjugated |
| Description | Mouse monoclonal antibody raised against a full length recombinant AQP8. |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | AQP8 |
| Gene Accession No. | BC040630 |
| Gene Symbols | AQP8 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.