Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AQP8, Mouse, Clone: 1F8, Abnova™

Catalog No. 89016175 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-175 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-175 Supplier Abnova Corporation Supplier No. H00000343M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant AQP8.

Aquaporin 8 (AQP8) is a water channel protein. Aquaporins are a family of small integral membrane proteins related to the major intrinsic protein (MIP or AQP0). Aquaporin 8 mRNA is found in pancreas and colon but not other tissues. [provided by RefSeq

Sequence: MCEPEFGNDKAREPSVGGRWRVSWYERFVQPCLVELLGSALFIFIGCLSVIENGTDTGLLQPALAHGLALGLVIATLGNISGGHFNPAVSLAAMLIGGLNLVMLLPYWVSQLLGGMLGAALAKAVSPEERFWNASGAAFVTVQEQGQVAGALVAEIILTTLLALAVCMGAINEKTKGPLAPFSIGFAVTVDILAGGPVSGGCMNPARAFGPAVVANHWNFHWIYWLGPLLAGLLVGLLIRCFIGDGKTRLILKAR

Specifications

Antigen AQP8
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 1F8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant AQP8.
Formulation PBS with no preservative; pH 7.4
Gene AQP8
Gene Accession No. BC040630
Gene Symbols AQP8
Host Species Mouse
Immunogen AQP8 (AAH40630, 1 a.a. ∼ 255 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 343
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.