Learn More
Description
Specifications
Specifications
| Antigen | Aquaporin 0 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in PBS, pH 7.4 with 0.1% BSA and 0.1% sodium azide. |
| Gene Symbols | MIP; Aquaporin-0; aquaporin; AQP0; LIM1; MP26; MIP26 |
| Host Species | Rabbit |
| Immunogen | A 17 AA synthetic peptide within the carboxy terminal domain of human Aquaporin-0 (Shiels et al. 1988; Kent et al. 1990; Pisano et al. 1991; Shiels et al. 1996) was selected for antibody production. This domain is predicted to be cytoplasmic. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.