Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 1 Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3272200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3272200UL 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3272200UL Supplier MilliporeSigma Supplier No. AB3272200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-Aquaporin 1 Antibody is validated for use in immunohistochemistry, western blotting for the detection of Aquaporin 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 0.025% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).
Gene Symbols AQP1; Aquaporin-CHIP; MGC26324; AQP-CHIP; CO; Aquaporin-1; CHIP28; AQP-1
Host Species Rabbit
Immunogen Highly purified peptide corresponding to residues 242-260 of human Aquaporin 1 (SwissProt accession number P29972).
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_198098.1
Target Species Human, Rat
Content And Storage Lyophilized: −20°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.