Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB3594200UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3594200UL 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3594200UL Supplier MilliporeSigma Supplier No. AB3594200UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Aquaporin 4 using this Anti-Aquaporin 4 Antibody, C-terminus validated for use in immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal lyophilized from buffer containing PBS, pH 7.4, containing 1% BSA, and 0.025% sodium azide. Reconstitute with 200μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min).
Gene Symbols AQP4; MIWC; WCH4; AQP-4; HMIWC2; MGC22454
Host Species Rabbit
Immunogen GST fusion protein with residues 249-323 of rat Aquaporin 4 (SwissProt accession number P47863).
Purification Method Immunoaffinity purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001650.4
Target Species Human, Mouse, Rat
Content And Storage -20°C in undiluted aliquotes for up to 6 months, Avoid Freeze/Thaw Cycles
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.