Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Aquaporin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB2218
Description
Anti-Aquaporin 4 Antibody is an antibody against Aquaporin 4 for use in western blotting.
Specifications
Aquaporin 4 | |
Polyclonal | |
Purified in PBS with 0.05% NaN3 | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Western Blot | |
Unconjugated | |
AQP4; MIWC; WCH4; AQP-4; aquaporin-4; HMIWC2; Aquaporin-4; MGC22454 | |
Synthetic Linear Peptide | |
100 μg | |
NM_001650.4; NM_004028.3 | |
2-8°C for 12 months in undiluted aliquotes |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction