Learn More
Aquaporin 4 Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma Products

Description
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Antigen | Aquaporin 4 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity Purified immunoglobulin. Lyophilized from PBS, pH 7.4, containing 1% BSA, and 0.025% sodium azide. Reconstitute with 50μL of sterile distilled water. Centrifuge antibody preparation before use (10,000 x g for 5 min). |
| Gene Symbols | AQP4; MIWC; WCH4; AQP-4; aquaporin-4; HMIWC2; Aquaporin-4; MGC22454 |
| Host Species | Rabbit |
| Immunogen | GST fusion protein with residues 249-323 of rat Aquaporin 4 (SwissProt accession number P47863). It is expected that the antibody may also react with mouse (73/75), bovine (71/75), human (69/75) and rabbit (64/75) due to sequence homology. |
| Purification Method | Affinity Purified |
| Quantity | 50 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.