Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Aquaporin 5, Polyclonal

Catalog No. 17861550UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
17-861-550UG 50 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-861-550UG Supplier MilliporeSigma™ Supplier No. 17861550UG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects Aquaporin 5 Clone: in Human, Mouse, Rat samples, and it is validated for ELISA, Immunofluorescence, Immunohistochemistry, Immuno Blotting

Rat AQP5 is a 265 amino acid transmembrane protein belonging to the aquaporin family. AQP5 is found in several epithelial tissues (salivary and lachrymal glands, corneal epithelium in the eye and lung). It is implicated in the generation of saliva, tears, and pulmonary secretions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 5
Applications ELISA, Immunoblot, Immunoblot, Immunofluorescence, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation In PBS, 0.1% BSA.
Host Species Rabbit
Immunogen a synthetic peptide corresponding to a 17-amino acid sequence in the cytoplasmic region of rat AQP5
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽28kDa AQP 5 protein in epithelial tissue extracts.
Target Species Human, Mouse, Rat
Content And Storage −20°C
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.