Learn More
Description
Specifications
Specifications
| Antigen | Aquaporin 7 |
| Applications | ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in 100mM Tris, pH 7.5 containing 0.2% BSA as stabilizer and 0.05% sodium azide. |
| Gene Symbols | AQP7; Aquaporin-7-like; AQP-7; AQPap; AQP9; MGC149555; Aquaporin-7; MGC149556; AQP7L |
| Host Species | Chicken |
| Immunogen | An 18 AA synthetic peptide within the N-terminal domain of rat Aquaporin-7 (Ishibashi et al. 1997) was selected for antibody production. This domain is predicted to be cytoplasmic. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.