Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aquaporin 7 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB1556850UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1556850UL 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1556850UL Supplier MilliporeSigma Supplier No. AB1556850UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Aquaporin 7 Antibody detects level of Aquaporin 7 and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aquaporin 7
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 50μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).
Gene Symbols AQP7; Aquaporin-7-like; AQP-7; AQPap; AQP9; MGC149555; Aquaporin-7; MGC149556; AQP7L
Host Species Rabbit
Immunogen Purified peptide from the N-terminus of rat AQP7 (Accession number P56403).
Purification Method Immunoaffinity purified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_001170.1
Target Species Mouse, Rat
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.