Learn More
AREG, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human AREG peptide using ARM Technology.
Supplier: Abnova Corporation H00000374K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
 - Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
 
Specifications
| AREG | |
| Monoclonal | |
| Unconjugated | |
| In 1X PBS, pH 7.4 | |
| AREG | |
| AREG | |
| A synthetic peptide of human AREG | |
| RUO | |
| 374 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG | 
| ELISA, Western Blot | |
| 1mg | |
| amphiregulin | |
| AREG | |
| AR, CRDGF, MGC13647, SDGF | |
| Rabbit | |
| 1 mg x up to 3 Abs | |
| Primary | |
| Human | |
| Liquid | 
Your input is important to us. Please complete this form to provide feedback related to the content on this product.