Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARF1 Mouse anti-Human, Mouse, Rat, Clone: 3F1 , MilliporeSigma™

Catalog No. 051427MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-142-7MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-142-7MI Supplier MilliporeSigma Supplier No. 051427
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-ARF1 Antibody, clone 3F1 detects level of ARF1 and has been published and validated for use in western blot, ELISA, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARF1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 3F1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2a in 0.02Mphosphate buffer, 0.25MNaCl, pH 7.6, with 0.1% sodium azide
Gene Accession No. NM_001024226.1
Gene Symbols ARF1
Host Species Mouse
Immunogen Synthetic peptide corresponding to the C-terminal amino acid sequence of human ARF1.
Purification Method Purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.