Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Arginase-1 Chicken anti-Mouse, Rat, Guinea Pig, Human, Polyclonal, MilliporeSigma™

Catalog No. ABS535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS535MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS535MI Supplier MilliporeSigma Supplier No. ABS535
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

This Anti-Arginase-1 Antibody is validated for use in Western Blotting and Immunohistochemistry for the detection of Arginase-1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Arginase-1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken Polyclonal other in buffer containing PBS with 0.05% sodium azide.
Gene Symbols ARG1
Host Species Chicken
Immunogen Full-length recombinant protein corresponding to Rat Arginase-1.
Purification Method IgY purified from Yolk
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_058830
Target Species Guinea Pig, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.