Learn More
Rho GTPase activating protein 4, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human ARHGAP4 protein.
Supplier: Abnova Corporation H00000393D01P
Description
This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of small GTP-binding proteins belonging to the RAS superfamily. The protein encoded by the orthologous gene in rat is localized to the Golgi complex and can redistribute to microtubules. The rat protein stimulates the activity of some Rho GTPases in vitro. Genomic deletions of this gene and a neighboring gene have been found in patients with nephrogenic diabetes insipidus. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: MAAHGKLRRERGLQAEYETQVKEMRWQLSEQLRCLELQGELRRELLQELAEFMRRRAEVELEYSRGLEKLAERFSSRGGRLGSSREHQSFRKEPSLLSPLHCWAVLLQHTRQQSRESAALSEVLAGPLAQRLSHIAEDVGRLVKKSRDLEQQLQDELLEVVSELQTAKKTYQAYHMESVNAEAKLREAERQEEKRAGRSVPTTTAGATEAGPLRKSSLKKGGRLVEKLWPPQRPVAASSCAPVCWLQAGFLVHPPWWGAMCAPSTHQRQAKFMEHKLKCTKARNEYLLSLASVNAAVSNYYLHDVLDLMDCCDTGFHLALGQVLRSYTAAESRTQASQVQGLGSLEEAVEALDPPGDKAKVLEVHATVFCPPLRFDYHPHDGDEVAEICVEMELRDEILPRAQNIQSRLDRQTIETEEVNKTLKATLQALLEVVASDDGDVLDSFQTSPSTESLKSTSSDPGSRQAGRRRGQQQETETFYLTKLQEYLSGRSILAKLQAKHEKLQEALQRGDKEEQEVSWTQYTQRKFQKSRQPRPSSQYNQRLFGGDMEKFIQSSGQPVPLVVESCIRFINLNGLQHEGIFRVSGAQLRVSEIRDAFERGEDPLVEGCTAHDLDSVAGVLKLYFRSLEPPLFPPDLFGELLASSELEATAERVEHVSRLLWRLPAPVLVVLRYLFTFLNHLAQYSDENMMDPYNLAVCFGPTLLPVPAGQDPVALQGRVNQLVQTLIVQPDRVFPPLTSLPGPVYEKCMAPPSASCLGDAQLESLGADNEPELEAEMPAQEDDLEGVVEAVACFAYTGRTAQELSFRRGDVLRLHERASSDWWRGEHNGMRGLIPHKYITLPAGTEKQVVGAGLQTAGESGSSPEGLLASELVHRPEPCTSPEAMGPSGHRRRCLVPASPEQHVEVDKAVAQNMDSVFKELLGKTSVRQGLGPASTTSPSPGPRSPKAPPSSRLGRNKGFSRGPGAPASPSASHPQGLDTTPKPHSpecifications
Rho GTPase activating protein 4 | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human ARHGAP4 protein. | |
ARHGAP4 | |
C1/KIAA0131/RGC1/RhoGAP4/p115 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC052303 | |
ARHGAP4 | |
ARHGAP4 (AAH52303.1, 1 a.a. ∼ 986 a.a) full-length human protein. | |
100 μg | |
Membrane Receptors | |
393 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.