Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARHGAP6 (M01A), Mouse anti-Human, Clone: 6B3, Abnova™

Catalog No. 89016209 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-209 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-209 Supplier Abnova Corporation Supplier No. H00000395M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ARHGAP6.

This gene encodes a member of the rhoGAP family of proteins which play a role in the regulation of actin polymerization at the plasma membrane during several cellular processes. This protein is thought to have two independent functions, one as a GTPase-activating protein with specificity for RhoA, and another as a cytoskeletal protein that promotes actin remodeling. Multiple alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: RHADDNISKDGQEVTGNKMTSLNLATIFGPNLLHKQKSSDKEFSVQSSARAEESTAIIAVVQKMIENYEALFMVPPDLQNEVLISLLETDPDVVDYLLRRKASQSS

Specifications

Antigen ARHGAP6
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ARHGAP6.
Formulation ascites with no preservative
Gene ARHGAP6
Gene Accession No. NM_013427
Gene Alias RHOGAP6/RHOGAPX-1
Gene Symbols ARHGAP6
Host Species Mouse
Immunogen ARHGAP6 (NP_038286, 531 a.a. ∼ 636 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 395
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.