Learn More
Rho GDP dissociation inhibitor (GDI) alpha (A02), Mouse anti-Human, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length recombinant ARHGDIA.
Supplier: Abnova Corporation H00000396A02
Description
Aplysia Ras-related homologs (ARHs), also called Rho genes, belong to the RAS gene superfamily encoding small guanine nucleotide exchange (GTP/GDP) factors. The ARH proteins may be kept in the inactive, GDP-bound state by interaction with GDP dissociation inhibitors, such as ARHGDIA (Leffers et al., 1993 [PubMed 8262133]).[supplied by OMIM
Sequence: MAEQEPTAEQLAQIAAENEEDEHSVNYKPPAQKSIQEIQELDKDDESLRKYKEALLGRVAVSADPNVPNVVVTGLTLVCSSAPGPLELDLTGDLESFKKQSFVLKEGVEYRIKISFRVNREIVSGMKYIQHTYRKGVKIDKTDYMVGSYGPRAEEYEFLTPVEEAPKGMLARGSYSIKSRFTDDDKTDHLSWEWNLTIKKDWKDSpecifications
Rho GDP dissociation inhibitor (GDI) alpha | |
Polyclonal | |
Mouse polyclonal antibody raised against a full-length recombinant ARHGDIA. | |
ARHGDIA | |
GDIA1/MGC117248/RHOGDI/RHOGDI-1 | |
Mouse | |
50 μL | |
Signal Transduction | |
Primary | |
Human | |
Serum |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
BC005851 | |
ARHGDIA | |
ARHGDIA (AAH05851, 1 a.a. ∼ 204 a.a) full-length recombinant protein with GST tag. | |
RUO | |
Yes | |
396 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.