Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho GDP dissociation inhibitor (GDI) gamma (A01), Mouse anti-Human, Polyclonal Antibody, Abnova™

Catalog No. 89004022 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-022 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-022 Supplier Abnova Corporation Supplier No. H00000398A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length recombinant ARHGDIG.

Sequence: MLGLDACELGAQLLELLRLALCARVLLADKEGGPPAVDEVLDEAVPEYRAPGRKSLLEIRQLDPDDRSLAKYKRVLLGPLPPAVDPSLPNVQVTRLTLLSEQAPGPVVMDLTGDLAVLKDQVFVLKEGVDYRVKISFKVHREIVSGLKCLHHTYRRGLRVDKTVYMVGSYGPSAQEYEFVTPVEEAPRGALVRGPYLVVSLFTDDDRTHHLSWEWGLCICQDWKD

Specifications

Antigen Rho GDP dissociation inhibitor (GDI) gamma
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length recombinant ARHGDIG.
Formulation 50% glycerol
Gene ARHGDIG
Gene Accession No. BC047699
Gene Alias RHOGDI-3
Gene Symbols ARHGDIG
Host Species Mouse
Immunogen ARHGDIG (AAH47699, 1 a.a. ∼ 225 a.a) full-length recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Signal Transduction
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 398
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.