Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho guanine nucleotide exchange factor (GEF) 3, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89017070 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-017-070 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-017-070 Supplier Abnova Corporation Supplier No. H00050650B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ARHGEF3 protein.

Rho-like GTPases are involved in a variety of cellular processes, and they are activated by binding GTP and inactivated by conversion of GTP to GDP by their intrinsic GTPase activity. Guanine nucleotide exchange factors (GEFs) accelerate the GTPase activity of Rho GTPases by catalyzing their release of bound GDP. This gene encodes a guanine nucleotide exchange factor, which specifically activates two members of the Rho GTPase family: RHOA and RHOB, both of which have a role in bone cell biology. It has been identified that genetic variation in this gene plays a role in the determination of bone mineral density (BMD), indicating the implication of this gene in postmenopausal osteoporosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MKLPCLSSYDSYCSNQVAAKALLDHKKQDHRVQDFLQRCLESPFSRKLDLWNFLDIPRSRLVKYPLLLREILRHTPNDNPDQQHLEEAINIIQGIVAEINTKTGESECRYYKERLLYLEEGQKDSLIDSSRVVCCHGELKNNRGVKLHVFLFQEVLVITRAVTHNEQLCYQLYRQPIPVKDLLLEDLQDGEVRLGGSLRGAFSNNERIKNFFRVSFKNGSQSQTHSLQANDTFNKQQWLNCIRQAKETVLCAAGQAGVLDSEGSFLNPTTGSRELQGETKLEQMDQSDSESDCSMDTSEVSLDCERMEQTDSSCGNSRHGESNV

Specifications

Antigen Rho guanine nucleotide exchange factor (GEF) 3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ARHGEF3 protein.
Formulation PBS with no preservative; pH 7.4
Gene ARHGEF3
Gene Accession No. BC099715
Gene Alias DKFZp434F2429/FLJ98126/GEF3/MGC118905/STA3/XPLN
Gene Symbols ARHGEF3
Host Species Mouse
Immunogen ARHGEF3 (AAH99715, 1 a.a. ∼ 324 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 50650
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.