Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rho guanine nucleotide exchange factor (GEF) 5, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89006527 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-006-527 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-006-527 Supplier Abnova Corporation Supplier No. H00007984B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human ARHGEF5 protein.

Rho GTPases play a fundamental role in numerous cellular processes initiated by extracellular stimuli that work through G protein coupled receptors. The encoded protein may form a complex with G proteins and stimulate Rho-dependent signals. This protein may be involved in the control of cytoskeletal organization. [provided by RefSeq

Sequence: MGGFSRRCSKLINSSQLLYQEYSDVVLNKEIQSQQRLESLSETPGPSSPRQPRKALVSSESYLQRLSMASSGSLWQEIPVVRNSTVLLSMTHEDQKLQEVKFELIVSEASYLRSLNIAVDHFQLSTSLRATLSNQEHQWLFSRLQDVRDVSATFLSDLEENFENNIFSFQVCDVVLNHAPDFRRVYLPYVTNQTYQERTFQSLMNSNSNFREVLEKLESDPVCQRLSLKSFLILPFQRITRLKLLLQNILKRTQPGSSEEAEATKAHHALEQLIRDCNNNVQSMRRTEELIYLSQKIEFECKIFPLISQSRWLVKSGELTALEFSASPGLRRKLNTRPVHLHLFNDCLLLSRPREGSRFLVFDHAPFSSIRGEKCEMKLHGPHKNLFRLFLRQNTQGAQAEFLFRTETQSEKLRWISALAMPREELDLLECYNSPQVQCLRAYKPRENDELALEKADVVMVTQQSSDGWLEGVRLSDGERGWFPVQQVEFISNPEVRAQNLKEAHRVKTAKLQLVEQQA

Specifications

Antigen Rho guanine nucleotide exchange factor (GEF) 5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human ARHGEF5 protein.
Formulation No additive
Gene ARHGEF5
Gene Accession No. Q12774
Gene Alias DKFZp686N1969/GEF5/P60/TIM/TIM1
Gene Symbols ARHGEF5
Host Species Mouse
Immunogen ARHGEF5 (Q12774, 1 a.a. ∼ 519 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 7984
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.