Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Rat ARK5/NUAK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6978
This item is not returnable.
View return policy
Description
ARK5/NUAK1 Monoclonal specifically detects ARK5/NUAK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
ARK5/NUAK1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
O60285 | |
NUAK1 | |
E. coli-derived recombinant human ARK5/NUAK1 Lys311-Met462 Accession # O60285 | |
100 μg | |
Primary | |
Detects human ARK5/NUAK1 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human SNARK is observed. | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
715901 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
AMPK-related protein kinase 5, ARK5AMP-activated protein kinase family member 5, EC 2.7.11, EC 2.7.11.1, KIAA0537NUAK family SNF1-like kinase 1, NUAK family, SNF1-like kinase, 1, NuaK1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
9891 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction