Learn More
ARL2BP, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human ARL2BP peptide using ARM Technology.
Supplier: Abnova Corporation H00023568K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
ADP-ribosylation factor (ARF)-like proteins (ARLs) comprise a functionally distinct group of the ARF family of RAS-related GTPases. The protein encoded by this gene binds to ARL2.GTP with high affinity but does not interact with ARL2.GDP, activated ARF, or RHO proteins. The lack of detectable membrane association of this protein or ARL2 upon activation of ARL2 is suggestive of actions distinct from those of the ARFs. This protein is considered to be the first ARL2-specific effector identified, due to its interaction with ARL2.GTP but lack of ARL2 GTPase-activating protein activity. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
ARL2BP | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
ARL2BP | |
ARL2BP | |
A synthetic peptide of human ARL2BP | |
RUO | |
23568 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
ADP-ribosylation factor-like 2 binding protein | |
ARL2BP | |
BART, BART1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.