Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (2E7), Biotin, Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Supplier: Novus Biologicals NBP128125
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, In vitro assay.Specifications
Integrin alpha E/CD103 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, In vitro assay | |
ITGAE | |
C57BL/6J mouse intestinal intraepithelial lymphocytes (IEL) | |
0.25 mg | |
Innate Immunity | |
3682 | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), In vitro Assay | |
20000000 | |
Biotin | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Armenian Hamster | |
Protein A or G purified | |
RUO | |
Primary | |
The CD103/alpha IELbeta7 integrin complex (Mr 160 and 115 kDa peptides). | |
Store at 4C. Do not freeze. | |
IgG2a |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction