Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MANF Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP132037
Description
MANF Polyclonal specifically detects MANF in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
ARMET | |
Polyclonal | |
Unconjugated | |
PBS (pH 7.0) and 40% Glycerol with 0.01% Thimerosal | |
MANF | |
Recombinant protein encompassing a sequence within the center region of human ARMET. The exact sequence is proprietary. | |
Affinity Purified | |
RUO | |
Primary | |
Tested in human. Predicted cross-reactivity based on sequence identitiy: Zebrafish (84%) and Xenopus tropicalis (89%). Not yet tested in any other species. | |
Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
1.0mg/mL | |
Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000 | |
arginine-rich, mutated in early stage tumors, ARMETArginine-rich protein, ARPMGC142150, mesencephalic astrocyte-derived neurotrophic factor, MGC142148, Protein ARMET | |
Rabbit | |
21 kDa | |
100 μL | |
Cancer, DNA Repair | |
7873 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction