Learn More
Description
Specifications
Specifications
| Antigen | Arp2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Affinity purified immunoglobulin. Liquid in 0.02M phosphate buffer, 0.25M NaCl, with 0.1% sodium azide as a preservative. |
| Gene Symbols | ACTR2; ARP2 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids 320-341 of human Arp2. The immunogen sequence is 100% identical in mouse, rat, Xenopus, and chicken. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.