Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-ARSB Clone: 1A4,

Catalog No. AP1139100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
AP1139100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP1139100UG Supplier MilliporeSigma™ Supplier No. AP1139100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ARSB Clone: 1A4 in Human samples, and it is validated for Immuno Blotting

Arylsulfatase B (ARSB) belongs to the sulfatase family. The arylsulfatase B homodimer catalyzes the hydrolysis of the sulfate groups of N-Acetyl-D-galactosamine, chondriotin sulfate, and dermatan sulfate. Defects in this gene cause Maroteaux-Lamy syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ARSB
Applications Immunoblot
Classification Monoclonal
Clone 1A4
Conjugate Unconjugated
Formulation In PBS, pH 7.2
Host Species Mouse
Immunogen A recombinant polypeptide corresponding to amino acids of 166-266 human ARSB, expressed as a GST fusion protein
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ARSB protein.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.