Learn More
ADP-ribosyltransferase 3, Mouse, Clone: 3A2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant ART3.
Supplier: Abnova Corporation H00000419M05
Description
ADP-ribosylation is a reversible posttranslational modification used to regulate protein function. ADP-ribosyltransferases, such as ART3, transfer ADP-ribose from NAD+ to the target protein, and ADP-ribosylhydrolases (see PARG; MIM 603501) reverse the reaction (Glowacki et al., 2002 [PubMed 12070318]).[supplied by OMIM
Sequence: LDMADNAFDDEYLKCTDRMEIKYVPQLLKEEKASHQQLDTVWENAKAKWAARKTQIFLPMNFKDNHGIALMAYISEAQEQTPFYHLFSEAVKMAGQSREDYIYGFQFKAFSpecifications
ADP-ribosyltransferase 3 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001179 | |
ART3 | |
ART3 (NP_001170, 29 a.a. ∼ 138 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
3A2 | |
Mouse monoclonal antibody raised against a partial recombinant ART3. | |
ART3 | |
FLJ26404 | |
Mouse | |
Affinity Purified | |
RUO | |
419 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.