Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARVCF, Mouse, Clone: 5D2, Abnova™

Catalog No. 89016219 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-219 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-219 Supplier Abnova Corporation Supplier No. H00000421M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ARVCF.

Armadillo Repeat gene deleted in Velo-Cardio-Facial syndrome (ARVCF) is a member of the catenin family which play an important role in the formation of adherens junction complexes, which are thought to facilitate communication between the inside and outside environments of a cell. ARVCF gene was isolated in the search for the genetic defect responsible for the autosomal dominant Velo-Cardio-Facial syndrome (VCFS) a relatively common human disorder with phenotypic features including cleft palate, conotruncal heart defects and facial dysmorphology. ARVCF gene encodes a protein containing two motifs, a coiled coil domain in the N-terminus and a 10 armadillo repeat sequence in the midregion. Since these sequences can facilitate protein-protein interactions ARVCF is thought to function in a protein complex. In addition, ARVCF contains a predicted nuclear-targeting sequence suggesting that it may have a function as a nuclear protein. [provided by RefSeq

Sequence: LSPGGFDDSTLPLVDKSLEGEKTGSRDVIPMDALGPDGYSTVDRRERRPRGASSAGEASEKEPLKLDPSRKAPPPGPSRPAVRLVDAVGDAKPQPVDSWV

Specifications

Antigen ARVCF
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 5D2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ARVCF.
Formulation PBS with no preservative; pH 7.4
Gene ARVCF
Gene Accession No. NM_001670
Gene Alias FLJ35345
Gene Symbols ARVCF
Host Species Mouse
Immunogen ARVCF (NP_001661, 863 a.a. ∼ 962 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 421
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.