Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AS160 Rabbit anti-Mouse, Human, Canine, Rat, Polyclonal, MilliporeSigma™

Catalog No. ABS54MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABS54MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABS54MI Supplier MilliporeSigma Supplier No. ABS54
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-AS160 Antibody detects level of AS160 and has been published and validated for use in western blotting and immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AS160
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4, 150mM NaCl) with 0.05% sodium azide.
Gene Symbols AS160
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of mouse AS160.
Purification Method Affinity Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001074747
Target Species Canine, Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.